Skip to Content
MilliporeSigma
Tributyl phosphate for extraction analysis, ≥99.0% (GC)

Tributyl phosphate

Tributyl phosphate for extraction analysis, ≥99.0% (GC)
Synonym(s):
TBP, TBPA, Tributyl phosphate
Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique