Skip to Content
MilliporeSigma
Tributylhexadecylphosphonium bromide 97%

Tributylhexadecylphosphonium bromide

Tributylhexadecylphosphonium bromide 97%
Synonym(s):
Hexadecyltributylphosphonium bromide, TBHDPB
Linear Formula:
CH3(CH2)15P+[(CH2)3CH3]3Br-
CAS Number:
Molecular Weight:
507.65
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing