Skip to Content
MilliporeSigma
Triafamone PESTANAL®, analytical standard

Triafamone

Triafamone PESTANAL®, analytical standard
Synonym(s):
2′-[(4,6-Dimethoxy-1,3,5-triazin-2-yl)carbonyl]-1,1,6′-trifluoro-N-methylmethanesulfonanilide, N-[2-[(4,6-Dimethoxy-1,3,5-triazin-2-yl)carbonyl]-6-fluorophenyl]-1,1-difluoro-N-methylmethanesulfonamide
Empirical Formula (Hill Notation):
C14H13F3N4O5S
CAS Number:
Molecular Weight:
406.34

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing