![Tranylcypromine Sulfate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/226/056/4a62fe41-7f8e-4045-ba39-c0a2f62c238e/640/4a62fe41-7f8e-4045-ba39-c0a2f62c238e.png)
Tranylcypromine sulfate
![Tranylcypromine Sulfate pharmaceutical secondary standard, certified reference material](/deepweb/assets/sigmaaldrich/product/structures/226/056/4a62fe41-7f8e-4045-ba39-c0a2f62c238e/640/4a62fe41-7f8e-4045-ba39-c0a2f62c238e.png)
Synonym(s):
trans-2-Phenylcyclopropylamine hemisulfate salt, Tranylcypromine
Linear Formula:
C9H11N · 1/2H2SO4
CAS Number:
Molecular Weight:
182.23
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2