Skip to Content
MilliporeSigma
trans,trans-Dibenzylideneacetone 98%

trans,trans-Dibenzylideneacetone

trans,trans-Dibenzylideneacetone 98%
Synonym(s):
1,5-Diphenyl-1,4-pentadien-3-one, trans,trans-1,5-Diphenyl-1,4-pentadien-3-one
Linear Formula:
C6H5CH=CHCOCH=CHC6H5
CAS Number:
Molecular Weight:
234.29

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing