Skip to Content
MilliporeSigma
trans,trans-2,4-Hexadienoic acid potassium salt ReagentPlus®, 99%

trans,trans-2,4-Hexadienoic acid potassium salt

trans,trans-2,4-Hexadienoic acid potassium salt ReagentPlus®, 99%
Synonym(s):
Potassium sorbate, Potassium 2,4-hexadienoate, Sorbic acid potassium salt
Linear Formula:
CH3CH=CHCH=CHCOOK
CAS Number:
Molecular Weight:
150.22
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing