trans,trans-2,4-Hexadien-1-ol
Synonym(s):
Sorbic alcohol
Linear Formula:
CH3CH=CHCH=CHCH2OH
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing