Skip to Content
MilliporeSigma
trans-3-(Trimethylsilyl)allyl alcohol 96%

trans-3-(Trimethylsilyl)allyl alcohol

trans-3-(Trimethylsilyl)allyl alcohol 96%
Synonym(s):
trans-(3-Hydroxy-2-propen-1-yl)trimethylsilane, trans-3-(Trimethylsilyl)-2-propen-1-ol
Linear Formula:
(CH3)3SiCH=CHCH2OH
CAS Number:
Molecular Weight:
130.26

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing