Skip to Content
MilliporeSigma
trans-3-Hexenoic acid 97%

trans-3-Hexenoic acid

trans-3-Hexenoic acid 97%
Linear Formula:
C2H5CH=CHCH2CO2H
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing