Skip to Content
MilliporeSigma
trans-2-Hexenyl isovalerate ≥95%, FG

trans-2-Hexenyl isovalerate

trans-2-Hexenyl isovalerate ≥95%, FG
Synonym(s):
2-Hexenyl 3-methylbutanoate
Empirical Formula (Hill Notation):
C11H20O2
CAS Number:
Molecular Weight:
184.28

Products

Product Category
Brand
Formula Weight
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing