Skip to Content
MilliporeSigma
trans-1,2-Bis(phenylsulfonyl)ethylene 98%

trans-1,2-Bis(phenylsulfonyl)ethylene

trans-1,2-Bis(phenylsulfonyl)ethylene 98%
Linear Formula:
C6H5SO2CH=CHSO2C6H5
CAS Number:
Molecular Weight:
308.37

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing