Skip to Content
MilliporeSigma
TOYOPEARL® AF-Red-650M bottle of 25 mL, 65 μm particle size

TOYOPEARL® AF-Red-650M

TOYOPEARL® AF-Red-650M bottle of 25 mL, 65 μm particle size
Synonym(s):
TOYOPEARL® AF-Red-650M Bulk Media

Products

Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing