Tomatidine hydrochloride
Synonym(s):
3β-Hydroxy-5α-tomatidane hydrochloride, 5α-Tomatidan-3β-ol hydrochloride
Empirical Formula (Hill Notation):
C27H45NO2 · HCl
CAS Number:
Molecular Weight:
452.11
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing