Tizanidine Related Compound C
Synonym(s):
1-Acetyl-2-imidazolidinethione
Empirical Formula (Hill Notation):
C5H8N2OS
CAS Number:
Molecular Weight:
144.19
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2