Skip to Content
MilliporeSigma
Titanium(IV) chloride solution 0.09 M in 20% HCl

Titanium(IV) chloride solution

Titanium(IV) chloride solution 0.09 M in 20% HCl
Synonym(s):
Titanium tetrachloride
Linear Formula:
TiCl4
CAS Number:
Molecular Weight:
189.68

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core
Showing 1-2 of 2