
Tin(II) chloride dihydrate

Synonym(s):
Stannous chloride dihydrate, Tin(II) chloride dihydrate
Linear Formula:
SnCl2 · 2H2O
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-10 of 10
Compare
Product Number
Description
Pricing