Skip to Content
MilliporeSigma
Timolol maleate British Pharmacopoeia (BP) Reference Standard

Timolol maleate

Timolol maleate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Timolol maleate salt, S-(−)-1-(t-Butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-2-propanol maleate salt, Timolol hydrogen maleate
Empirical Formula (Hill Notation):
C13H24N4O3S · C4H4O4
CAS Number:
Molecular Weight:
432.49
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing