Skip to Content
MilliporeSigma
Tiglic aldehyde ≥96%

Tiglic aldehyde

Tiglic aldehyde ≥96%
Synonym(s):
(E)-2-Methylbut-2-enal, trans-2,3-Dimethylacrolein, trans-2-Methyl-2-butenal, Tiglinaldehyde
Linear Formula:
CH3CH=C(CH3)CHO
CAS Number:
Molecular Weight:
84.12

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing