Skip to Content
MilliporeSigma
Thyrotropin releasing hormone ≥98% (HPLC), powder

Thyrotropin releasing hormone

Thyrotropin releasing hormone ≥98% (HPLC), powder
Synonym(s):
L-Pyroglutamyl-L-histidyl-L-prolinamide, TRF, TRH, Thyroliberin, pGlu-His-Pro amide
Empirical Formula (Hill Notation):
C16H22N6O4
CAS Number:
Molecular Weight:
362.38
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing