Skip to Content
MilliporeSigma
Thymidine 5′-diphosphate sodium salt ≥96% (HPLC)

Thymidine 5′-diphosphate sodium salt

Thymidine 5′-diphosphate sodium salt ≥96% (HPLC)
Empirical Formula (Hill Notation):
C10H16N2O11P2 · xNa+
CAS Number:
Molecular Weight:
402.19 (free acid basis)

Products

Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing