Skip to Content
MilliporeSigma
Thiosalicylic acid 97%

Thiosalicylic acid

Thiosalicylic acid 97%
Synonym(s):
2-Mercaptobenzoic acid, Thiosalicylic acid
Linear Formula:
HSC6H4CO2H
CAS Number:
Molecular Weight:
154.19

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing