Skip to Content
MilliporeSigma
Thiophene analytical standard

Thiophene

Thiophene analytical standard
Synonym(s):
Thiofuran, Thiole, Thiophene
Empirical Formula (Hill Notation):
C4H4S
CAS Number:
Molecular Weight:
84.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing