Skip to Content
MilliporeSigma
Thiabendazole Pharmaceutical Secondary Standard; Certified Reference Material

Thiabendazole

Thiabendazole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
2-(4-Thiazolyl)benzimidazole, Thiabendazole
Empirical Formula (Hill Notation):
C10H7N3S
CAS Number:
Molecular Weight:
201.25
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Activity Spectrum
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Mode of Action
API Family
Reference Material Type
Special Grade
Technique