Skip to Content
MilliporeSigma
Thallium(I) cyclopentadienide 97%

Thallium(I) cyclopentadienide

Thallium(I) cyclopentadienide 97%
Synonym(s):
5-Cyclopentadienyl)thallium, Cyclopentadienylthallium, NSC 114719, Thallous cyclopentadienide
Empirical Formula (Hill Notation):
C5H5Tl
CAS Number:
Molecular Weight:
269.48
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing