Skip to Content
MilliporeSigma
Thalidomide pharmaceutical secondary standard, certified reference material

Thalidomide

Thalidomide pharmaceutical secondary standard, certified reference material
Synonym(s):
(±)-Thalidomide, (±)-2-(2,6-Dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione
Empirical Formula (Hill Notation):
C13H10N2O4
CAS Number:
Molecular Weight:
258.23

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade