Skip to Content
MilliporeSigma
Tetrakis(triphenylphosphine)palladium(0) 99%

Tetrakis(triphenylphosphine)palladium(0)

Tetrakis(triphenylphosphine)palladium(0) 99%
Synonym(s):
Palladium-tetrakis(triphenylphosphine), Pd(PPh3)4, Tetrakis(triphenylphosphine)-palladium(0)
Linear Formula:
Pd(P(C6H5)3)4
CAS Number:
Molecular Weight:
1155.56

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core