Skip to Content
MilliporeSigma
Tetrahydro-4-methyl-2-(2-methyl-1-propenyl)-2H-pyran mixture of cis and trans, ≥98%, stabilized, FG

Tetrahydro-4-methyl-2-(2-methyl-1-propenyl)-2H-pyran

Tetrahydro-4-methyl-2-(2-methyl-1-propenyl)-2H-pyran mixture of cis and trans, ≥98%, stabilized, FG
Synonym(s):
(+)-Rose oxide, (4S)-2-(2-Methyl-1-propenyl)-4-methyltetrahydropyran
Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1