Skip to Content
MilliporeSigma
Tetrahexylammonium hydroxide solution ~40% in H2O (T)

Tetrahexylammonium hydroxide solution

Tetrahexylammonium hydroxide solution ~40% in H2O (T)
Linear Formula:
[CH3(CH2)5]4N(OH)
CAS Number:
Molecular Weight:
371.68

Products

Product Category
Formula Weight
Physical Form
Functional Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing