Skip to Content
MilliporeSigma
Tetrafluoroboric acid diethyl ether complex

Tetrafluoroboric acid diethyl ether complex

Tetrafluoroboric acid diethyl ether complex
Synonym(s):
Fluoroboric acid diethyl ether complex
Linear Formula:
HBF4 · O(CH2CH3)2
CAS Number:
Molecular Weight:
161.93
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing