Skip to Content
MilliporeSigma
Tetraethylenepentamine pentahydrochloride 98%

Tetraethylenepentamine pentahydrochloride

Tetraethylenepentamine pentahydrochloride 98%
Synonym(s):
TEPA
Linear Formula:
HN(CH2CH2NHCH2CH2NH2)2·5HCl
CAS Number:
Molecular Weight:
371.61

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing