Skip to Content
MilliporeSigma
Tetrachlorophthalic anhydride 96%

Tetrachlorophthalic anhydride

Tetrachlorophthalic anhydride 96%
Synonym(s):
Tetrachlorophthalic anhydride
Empirical Formula (Hill Notation):
C8Cl4O3
CAS Number:
Molecular Weight:
285.90

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing