![Tetracaine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/174/627/ce9e3117-4684-4da3-8413-b7e686dca3a5/640/ce9e3117-4684-4da3-8413-b7e686dca3a5.jpg)
Tetracaine Related Compound C
![Tetracaine Related Compound C Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/174/627/ce9e3117-4684-4da3-8413-b7e686dca3a5/640/ce9e3117-4684-4da3-8413-b7e686dca3a5.jpg)
Synonym(s):
Methyl 4-(butylamino)benzoate, 4-(Butylamino)benzoic acid methyl ester
Empirical Formula (Hill Notation):
C12H17NO2
CAS Number:
Molecular Weight:
207.27
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2