Skip to Content
MilliporeSigma
Tetrabutylammonium hydrogen difluoride solution ~50% in acetonitrile

Tetrabutylammonium hydrogen difluoride solution

Tetrabutylammonium hydrogen difluoride solution ~50% in acetonitrile
Synonym(s):
TBABF, Tetrabutylammonium bifluoride, Tetrabutylammonium difluoride
Linear Formula:
(CH3CH2CH2CH2)4N(HF2)
CAS Number:
Molecular Weight:
281.47

Products

Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing