Skip to Content
MilliporeSigma
TES sodium salt ≥99% (titration)

TES sodium salt

TES sodium salt ≥99% (titration)
Synonym(s):
N-[Tris(hydroxymethyl)methyl]-2-aminoethanesulfonic acid sodium salt
Empirical Formula (Hill Notation):
C6H14NNaO6S
CAS Number:
Molecular Weight:
251.23

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing