Skip to Content
MilliporeSigma
TES-ADT >99% (HPLC)

TES-ADT

TES-ADT >99% (HPLC)
Synonym(s):
5,11-Bis(triethylsilylethynyl)anthradithiophene
Empirical Formula (Hill Notation):
C34H38S2Si2
CAS Number:
Molecular Weight:
566.97

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing