Skip to Content
MilliporeSigma
tert-Butyldimethylsilanol 99%

tert-Butyldimethylsilanol

tert-Butyldimethylsilanol 99%
Linear Formula:
(CH3)3CSi(CH3)2OH
CAS Number:
Molecular Weight:
132.28

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing