Skip to Content
MilliporeSigma
tert-Butyl acetoacetate reagent grade, 98%

tert-Butyl acetoacetate

tert-Butyl acetoacetate reagent grade, 98%
Linear Formula:
CH3COCH2COOC(CH3)3
CAS Number:
Molecular Weight:
158.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing