Skip to Content
MilliporeSigma
vibrant-m

Terlipressin for NMR identification

vibrant-m
Synonym(s):
Terlipressin
Empirical Formula (Hill Notation):
C52H74N16O15S2
CAS Number:
Molecular Weight:
1227.37
EC Number:

Products

Formula Weight
Manufacturer
Format
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1