Skip to Content
MilliporeSigma
Tenofovir Disoproxil Related Compound A certified reference material, pharmaceutical secondary standard

Tenofovir disoproxil Related Compound A

Tenofovir Disoproxil Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
5-[[(1S)-2-(6-Amino-9H-purin-9-yl)-1-methylethoxy]methyl]-2,4,6,8-tetraoxa-5-phosphanonanedioic acid 1,9-bis(1-methylethyl) ester 5-oxide (E)-2-butenedioate (1:1)
Empirical Formula (Hill Notation):
C19H30N5O10P · C4H4O4
CAS Number:
Molecular Weight:
635.51

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade