![Temazepam Related Compound G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/312/160/3a2fd8a1-09c8-475a-b854-b3f2ae1abb45/640/3a2fd8a1-09c8-475a-b854-b3f2ae1abb45.jpg)
Temazepam Related Compound G
![Temazepam Related Compound G certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/312/160/3a2fd8a1-09c8-475a-b854-b3f2ae1abb45/640/3a2fd8a1-09c8-475a-b854-b3f2ae1abb45.jpg)
Synonym(s):
(3RS)-7-Chloro-1,4-dimethyl-5-phenyl-4,5-dihydro-1H-1,4-benzodiazepin-2,3-dione
Empirical Formula (Hill Notation):
C17H15ClN2O2
CAS Number:
Molecular Weight:
314.77
Products
Brand
Formula Weight
Manufacturer
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2