
Temazepam Related Compound F

Synonym(s):
(3RS)-7-Chloro-1-methyl-5-phenyl-4,5-dihydro-1H-1,4-benzodiazepine-2,3-dione
Empirical Formula (Hill Notation):
C16H13ClN2O2
CAS Number:
Molecular Weight:
300.74
Products
Brand
Formula Weight
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing