Skip to Content
MilliporeSigma
Telmisartan for system suitability European Pharmacopoeia (EP) Reference Standard

Telmisartan for system suitability

Telmisartan for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Telmisartan, 4′[(1,4′-Dimethyl-2′-propyl[2,6′-bi-1H-benzimidazol]-1′-yl)methyl][1,1′-biphenyl]-2-carboxylic acid, BIBR 277
Empirical Formula (Hill Notation):
C33H30N4O2
CAS Number:
Molecular Weight:
514.62

Products

Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1