Skip to Content
MilliporeSigma
Tebuthiuron PESTANAL®, analytical standard

Tebuthiuron

Tebuthiuron PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C9H16N4OS
CAS Number:
Molecular Weight:
228.31
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing