Skip to Content
MilliporeSigma
TDCPP PESTANAL®, analytical standard

TDCPP

TDCPP PESTANAL®, analytical standard
Synonym(s):
Tris(1,3-dichloro-2-propyl) phosphate, 1,3-Dichloro-2-propanol phosphate, Phosphoric acid tris(1,3-dichloro-2-propyl ester)
Empirical Formula (Hill Notation):
C9H15Cl6O4P
CAS Number:
Molecular Weight:
430.90
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing