Skip to Content
MilliporeSigma
(t-Bu)2PMe-Pd-G3 95%

(t-Bu)2PMe-Pd-G3

(t-Bu)2PMe-Pd-G3 95%
Synonym(s):
Methanesulfonato (di-tert-butyl) methylphosphino (2′-amino-1,1′-biphenyl-2-yl) palladium(II)
Empirical Formula (Hill Notation):
C22H34NO3PPdS
CAS Number:
Molecular Weight:
529.97

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing