Skip to Content
MilliporeSigma
TAPSO ≥99% (titration)

TAPSO

TAPSO ≥99% (titration)
Synonym(s):
2-Hydroxy-3-[tris(hydroxymethyl)methylamino]-1-propanesulfonic acid, N-[Tris(hydroxymethyl)methyl]-3-amino-2-hydroxypropanesulfonic acid
Empirical Formula (Hill Notation):
C7H17NO7S
CAS Number:
Molecular Weight:
259.28
EC Number:

Products

Product Category
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing