Skip to Content
MilliporeSigma
T2-Toxin solution 100 μg/mL in acetonitrile, analytical standard

T2-Toxin solution

T2-Toxin solution 100 μg/mL in acetonitrile, analytical standard
Synonym(s):
Epoxytrichothecene, Fusariotoxin T 2, Mycotoxin T 2
Empirical Formula (Hill Notation):
C24H34O9
CAS Number:
Molecular Weight:
466.52
EC Number:

Products

Product Category
Brand
Formula Weight
Format
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing