Skip to Content
MilliporeSigma
(−)-Syringaresinol 4-(2′′-apiosylglucoside) phyproof® Reference Substance

(−)-Syringaresinol 4-(2′′-apiosylglucoside)

(−)-Syringaresinol 4-(2′′-apiosylglucoside) phyproof® Reference Substance
Synonym(s):
(-)-Syringaresnol-4-O-β-D-apiofuranosyl-(1→2)-β-D-glucopyranoside
Empirical Formula (Hill Notation):
C33H44O17
CAS Number:
Molecular Weight:
712.69

Products

Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Reference Material Type
Special Grade
Showing 1-1 of 1