![Synephrine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/371/8ffa47d1-6ea1-444c-a172-af19faffda52/640/8ffa47d1-6ea1-444c-a172-af19faffda52.png)
Synephrine
![Synephrine United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/341/371/8ffa47d1-6ea1-444c-a172-af19faffda52/640/8ffa47d1-6ea1-444c-a172-af19faffda52.png)
Synonym(s):
1-(4-Hydroxyphenyl)-2-methylaminoethanol, 4-Hydroxy-α-(methylaminomethyl)benzyl alcohol
Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing