Skip to Content
MilliporeSigma
Surfactin from Bacillus subtilis, ≥98.0% (HPLC)

Surfactin

Surfactin from Bacillus subtilis, ≥98.0% (HPLC)
Empirical Formula (Hill Notation):
C53H93N7O13
CAS Number:
Molecular Weight:
1036.34

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing